Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194488.2 | NP_001181417.1 | 693 | 123 | ||
XM_015151628.2 | XP_015007114.1 | 1590 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |