Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCKAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084186.4 | XP_001084186.1 | 916 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Macaca mulatta |