Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194066.1 | NP_001180995.1 | 1021 | 130 | ||
XM_028853361.1 | XP_028709194.1 | 1075 | 130 | |||
GenBank mRNA | DQ297681.1 | - | 860 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |