Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028841962.1 | XP_028697795.1 | 108 | 147 | ||
XM_028841963.1 | XP_028697796.1 | 108 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Macaca mulatta |