Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261540.1 | NP_001248469.1 | 420 | 146 | ||
XM_028849113.1 | XP_028704946.1 | 576 | 146 | |||
XM_028849114.1 | XP_028704947.1 | 564 | 146 | |||
GenBank mRNA | JU319705.1 | - | 420 | 146 | ||
JU336038.1 | - | 408 | 146 | |||
JV567215.1 | - | 502 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |