Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCLAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104502.4 | XP_001104502.1 | 788 | 125 | ||
XM_015112036.2 | XP_014967522.1 | 949 | 125 | |||
XM_015112040.2 | XP_014967526.1 | 940 | 125 | |||
XM_028831805.1 | XP_028687638.1 | 2245 | 125 | |||
XM_028831806.1 | XP_028687639.1 | 1256 | 125 | |||
XM_028831807.1 | XP_028687640.1 | 1230 | 125 | |||
GenBank mRNA | JU326659.1 | - | 536 | 125 | ||
JV647302.1 | - | 724 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |