Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001109056.4 | XP_001109056.1 | 1453 | 63 | ||
XM_015136562.2 | XP_014992048.1 | 1605 | 63 | |||
XM_015136563.2 | XP_014992049.1 | 1639 | 63 | |||
XM_015136564.2 | XP_014992050.1 | 1609 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |