Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028840786.1 | XP_028696619.1 | 42 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |