Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTNNBIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260588.1 | NP_001247517.1 | 156 | 77 | ||
XM_015148591.2 | XP_015004077.2 | 230 | 77 | |||
XM_015148689.2 | XP_015004175.2 | 241 | 77 | |||
XM_028844607.1 | XP_028700440.1 | 104 | 77 | |||
GenBank mRNA | JU335550.1 | - | 127 | 77 | ||
JU476479.1 | - | 195 | 77 | |||
JV044733.1 | - | 156 | 77 | |||
JV551586.1 | - | 112 | 77 | |||
JV690430.1 | - | 226 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |