Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265733.1 | NP_001252662.1 | 285 | 150 | ||
GenBank mRNA | JU333708.1 | - | 150 | |||
JU474943.1 | - | 266 | 150 | |||
JV046595.1 | - | 285 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |