Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028847356.1 | XP_028703189.1 | 1434 | 97 | ||
XM_028847359.1 | XP_028703192.1 | 1243 | 97 | |||
XM_028847362.1 | XP_028703195.1 | 1269 | 97 | |||
GenBank mRNA | JV497506.1 | - | 178 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |