Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087095.4 | XP_001087095.1 | 1768 | 151 | ||
XM_015130428.2 | XP_014985914.1 | 1883 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |