Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105273.4 | XP_001105273.2 | 2628 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |