Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266469.1 | NP_001253398.1 | 262 | 143 | ||
GenBank mRNA | JU333380.1 | - | 262 | 143 | ||
JV460110.1 | - | 143 |
Gene Symbol: PLAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028826420.1 | XP_028682253.1 | 413 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |