Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194119.3 | NP_001181048.1 | 837 | 148 | ||
XM_001099323.4 | XP_001099323.1 | 886 | 148 | |||
XM_015120818.2 | XP_014976304.1 | 903 | 148 | |||
XM_015120819.2 | XP_014976305.1 | 1105 | 148 | |||
XM_015120820.2 | XP_014976306.1 | 805 | 148 | |||
XM_028837149.1 | XP_028692982.1 | 990 | 148 | |||
XM_028837150.1 | XP_028692983.1 | 980 | 148 | |||
GenBank mRNA | JU336650.1 | - | 789 | 148 | ||
JU473391.1 | - | 815 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |