Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001096190.3 | XP_001096190.2 | 1167 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |