Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261150.1 | NP_001248079.1 | 1669 | 140 | ||
XM_015130128.2 | XP_014985614.1 | 2028 | 140 | |||
XM_015130129.2 | XP_014985615.1 | 2034 | 140 | |||
XM_015130130.2 | XP_014985616.1 | 1975 | 140 | |||
XM_015130131.2 | XP_014985617.1 | 1597 | 140 | |||
XM_015130132.2 | XP_014985618.1 | 1520 | 140 | |||
XM_015130133.2 | XP_014985619.1 | 2138 | 140 | |||
XM_028843187.1 | XP_028699020.1 | 2180 | 140 | |||
XM_028843188.1 | XP_028699021.1 | 1774 | 140 | |||
XM_028843189.1 | XP_028699022.1 | 1286 | 140 | |||
XM_028843191.1 | XP_028699024.1 | 1343 | 140 | |||
XM_028843192.1 | XP_028699025.1 | 2153 | 140 | |||
XM_028843193.1 | XP_028699026.1 | 1708 | 140 | |||
GenBank mRNA | JU320319.1 | - | 1933 | 140 | ||
JU320320.1 | - | 1933 | 140 | |||
JU473049.1 | - | 1833 | 140 | |||
JU473050.1 | - | 1833 | 140 | |||
JV044513.1 | - | 1669 | 140 | |||
JV044514.1 | - | 1669 | 140 | |||
JV645848.1 | - | 1936 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |