Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105980.4 | XP_001105980.3 | 668 | 89 | ||
XM_002802647.3 | XP_002802693.3 | 471 | 89 | |||
XM_002802648.3 | XP_002802694.3 | 535 | 89 | |||
XM_015129867.2 | XP_014985353.2 | 453 | 89 | |||
XM_015129869.2 | XP_014985355.2 | 459 | 89 | |||
GenBank mRNA | JU325604.1 | - | 386 | 89 | ||
JV057220.1 | - | 527 | 89 |