Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001094239.4 | XP_001094239.1 | 1059 | 137 | ||
GenBank mRNA | JU327448.1 | - | 873 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta |