Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028841375.1 | XP_028697208.1 | 1711 | 95 | ||
XM_028841376.1 | XP_028697209.1 | 1711 | 95 | |||
XM_028841383.1 | XP_028697216.1 | 1492 | 95 | |||
GenBank mRNA | JV706678.1 | - | 454 | 95 | ||
JV706679.1 | - | 454 | 95 |