Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257486.1 | NP_001244415.1 | 401 | 78 | ||
XM_015113535.2 | XP_014969021.1 | 398 | 78 | |||
XM_015113536.2 | XP_014969022.1 | 209 | 78 | |||
GenBank mRNA | JU337402.1 | - | 401 | 78 | ||
JV048868.1 | - | 590 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |