Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123353.1 | NP_001116825.1 | 497 | 124 | ||
XM_015115021.2 | XP_014970507.1 | 1194 | 124 | |||
XM_015115022.1 | XP_014970508.1 | 1634 | 124 | |||
XM_015115023.1 | XP_014970509.1 | 1746 | 124 | |||
GenBank mRNA | EU564533.1 | - | 497 | 124 | ||
JU478487.1 | - | 669 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |