Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015134138.2 | XP_014989624.1 | 495 | 161 | ||
XM_015134139.2 | XP_014989625.1 | 496 | 161 | |||
XM_028846088.1 | XP_028701921.1 | 495 | 161 | |||
XM_028846089.1 | XP_028701922.1 | 496 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |