Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8H8orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144763.2 | XP_015000249.1 | 276 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |