Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260971.2 | NP_001247900.1 | 265 | 64 | ||
XM_015137912.2 | XP_014993398.1 | 712 | 64 | |||
XM_015137913.2 | XP_014993399.1 | 1059 | 64 | |||
GenBank mRNA | JU473377.1 | - | 360 | 64 | ||
JV046759.1 | - | 267 | 64 | |||
JV464767.1 | - | 371 | 64 | |||
JV578416.1 | - | 200 | 64 |
Gene Symbol: STX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU341815.1 | - | 309 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |