Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194235.2 | NP_001181164.1 | 879 | 112 | ||
XM_015146359.2 | XP_015001845.1 | 1194 | 112 | |||
XM_015146360.2 | XP_015001846.1 | 1079 | 112 | |||
XM_015146362.2 | XP_015001848.1 | 1026 | 112 | |||
XM_015146363.2 | XP_015001849.1 | 796 | 112 | |||
XM_028853468.1 | XP_028709301.1 | 881 | 112 | |||
XM_028853469.1 | XP_028709302.1 | 768 | 112 | |||
GenBank mRNA | JU325905.1 | - | 379 | 112 | ||
JV662895.1 | - | 843 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |