Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261538.1 | NP_001248467.1 | 264 | 155 | ||
XM_015111245.2 | XP_014966731.2 | 1239 | 155 | |||
GenBank mRNA | JU328138.1 | - | 152 | 155 | ||
JU475966.1 | - | 264 | 155 | |||
JV716742.1 | - | 293 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |