Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AADAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001106694.4 | XP_001106694.1 | 292 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |