Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258145.1 | NP_001245074.1 | 907 | 82 | ||
GenBank mRNA | JU475159.1 | - | 907 | 82 | ||
JU475160.1 | - | 907 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |