Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135797.1 | NP_001129269.1 | 1011 | 133 | ||
XM_015124402.2 | XP_014979888.2 | 1280 | 133 | |||
XM_015124404.2 | XP_014979890.2 | 1276 | 133 | |||
XM_015124405.2 | XP_014979891.2 | 1195 | 133 | |||
XM_015124406.2 | XP_014979892.2 | 1021 | 133 | |||
XM_015124407.2 | XP_014979893.2 | 1286 | 133 | |||
XM_028838507.1 | XP_028694340.1 | 1301 | 133 | |||
XM_028838508.1 | XP_028694341.1 | 1267 | 133 | |||
XM_028838509.1 | XP_028694342.1 | 1282 | 133 | |||
XM_028838510.1 | XP_028694343.1 | 1298 | 133 | |||
XM_028838511.1 | XP_028694344.1 | 1319 | 133 | |||
XM_028838512.1 | XP_028694345.1 | 1315 | 133 | |||
XM_028838513.1 | XP_028694346.1 | 885 | 133 | |||
GenBank mRNA | EU204919.1 | - | 1011 | 133 | ||
JV600278.1 | - | 1181 | 133 | |||
JV650561.1 | - | 1218 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta Inventoried |