Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266558.1 | NP_001253487.1 | 382 | 128 | ||
XM_015126323.2 | XP_014981809.1 | 423 | 128 | |||
XM_028840379.1 | XP_028696212.1 | 423 | 128 | |||
XM_028840380.1 | XP_028696213.1 | 423 | 128 | |||
GenBank mRNA | JU336882.1 | - | 259 | 128 | ||
JU336883.1 | - | 259 | 128 | |||
JU472750.1 | - | 340 | 128 | |||
JU472751.1 | - | 340 | 128 | |||
JV043620.1 | - | 382 | 128 | |||
JV043621.1 | - | 382 | 128 | |||
JV664317.1 | - | 385 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |