Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015117450.1 | XP_014972936.1 | 569 | 100 | ||
XM_015117451.1 | XP_014972937.1 | 569 | 100 | |||
XM_015117452.1 | XP_014972938.1 | 569 | 100 | |||
XM_015117454.1 | XP_014972940.1 | 569 | 100 | |||
XM_015117455.1 | XP_014972941.1 | 569 | 100 | |||
GenBank mRNA | JU320602.1 | - | 533 | 100 | ||
JU324557.1 | - | 533 | 100 | |||
JU332867.1 | - | 533 | 100 | |||
JU332868.1 | - | 533 | 100 | |||
JU470478.1 | - | 499 | 100 | |||
JU470479.1 | - | 499 | 100 | |||
JV046159.1 | - | 407 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |