Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU333339.1 | - | 671 | 90 | ||
JV046423.1 | - | 623 | 90 | |||
JV046424.1 | - | 623 | 90 | |||
JV574083.1 | - | 686 | 90 | |||
JV574084.1 | - | 553 | 90 | |||
JV636019.1 | - | 610 | 90 | |||
JV636020.1 | - | 610 | 90 |