Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032837.1 | NP_001028009.1 | 139 | 155 | ||
GenBank mRNA | AF322912.1 | - | 139 | 155 | ||
JU475598.1 | - | 475 | 155 | |||
JU475599.1 | - | 475 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |