Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194606.1 | NP_001181535.1 | 243 | 59 | ||
XM_015111249.2 | XP_014966735.1 | 618 | 59 | |||
GenBank mRNA | JU334901.1 | - | 132 | 59 | ||
JU472104.1 | - | 174 | 59 | |||
JV045081.1 | - | 144 | 59 | |||
JV547387.1 | - | 170 | 59 | |||
JV584037.1 | - | 137 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |