Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266032.1 | NP_001252961.1 | 756 | 144 | ||
XM_028828927.1 | XP_028684760.1 | 2779 | 144 | |||
XM_028828928.1 | XP_028684761.1 | 702 | 144 | |||
XM_028828929.1 | XP_028684762.1 | 737 | 144 | |||
GenBank mRNA | JU320851.1 | - | 757 | 144 | ||
JU471484.1 | - | 725 | 144 | |||
JV045737.1 | - | 756 | 144 | |||
JV452056.1 | - | 752 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |