Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194028.1 | NP_001180957.1 | 352 | 119 | ||
GenBank mRNA | JU476476.1 | - | 347 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |