Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195652.1 | NP_001182581.1 | 639 | 137 | ||
GenBank mRNA | CO581132.1 | - | 643 | 137 | ||
JU326689.1 | - | 601 | 137 | |||
JU476699.1 | - | 691 | 137 | |||
JV044390.1 | - | 694 | 137 | |||
JV590057.1 | - | 661 | 137 | |||
JV658162.1 | - | 692 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |