Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042759.1 | NP_001036224.1 | 61 | 118 | ||
GenBank mRNA | U06694.1 | - | 61 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |