Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266152.1 | NP_001253081.1 | 362 | 111 | ||
XM_015135063.2 | XP_014990549.1 | 514 | 111 | |||
XM_015135064.2 | XP_014990550.1 | 583 | 111 | |||
XM_015135066.2 | XP_014990552.1 | 425 | 111 | |||
XM_015135068.2 | XP_014990554.1 | 514 | 111 | |||
XM_015135069.2 | XP_014990555.1 | 600 | 111 | |||
GenBank mRNA | JU322533.1 | - | 360 | 111 | ||
JU470814.1 | - | 362 | 111 | |||
JV045286.1 | - | 362 | 111 | |||
JV639470.1 | - | 365 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |