Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265729.1 | NP_001252658.1 | 2156 | 128 | ||
XM_015145733.2 | XP_015001219.1 | 2204 | 128 | |||
XM_015145734.2 | XP_015001220.1 | 2047 | 128 | |||
XM_028852118.1 | XP_028707951.1 | 1981 | 128 | |||
GenBank mRNA | JV636398.1 | - | 2156 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Macaca mulatta |