Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHLHE40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257518.1 | NP_001244447.1 | 515 | 85 | ||
GenBank mRNA | JU336853.1 | - | 515 | 85 | ||
JU477597.1 | - | 514 | 85 | |||
JV561297.1 | - | 565 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |