Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107617.4 | XP_001107617.1 | 638 | 146 | ||
XM_015145898.2 | XP_015001384.1 | 639 | 146 | |||
XM_015145905.2 | XP_015001391.1 | 658 | 146 | |||
XM_028831210.1 | XP_028687043.1 | 677 | 146 | |||
GenBank mRNA | JU321232.1 | - | 589 | 146 | ||
JU473295.1 | - | 599 | 146 | |||
JV048550.1 | - | 513 | 146 | |||
JV452729.1 | - | 612 | 146 | |||
JV489142.1 | - | 582 | 146 | |||
JV704658.1 | - | 623 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |