Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107617.4 | XP_001107617.1 | 688 | 95 | ||
XM_015145898.2 | XP_015001384.1 | 689 | 95 | |||
XM_015145905.2 | XP_015001391.1 | 708 | 95 | |||
XM_028831210.1 | XP_028687043.1 | 727 | 95 | |||
GenBank mRNA | JU321232.1 | - | 639 | 95 | ||
JU473295.1 | - | 649 | 95 | |||
JV048550.1 | - | 563 | 95 | |||
JV452729.1 | - | 662 | 95 | |||
JV489142.1 | - | 632 | 95 | |||
JV704658.1 | - | 673 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |