Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107617.4 | XP_001107617.1 | 836 | 85 | ||
XM_015145898.2 | XP_015001384.1 | 837 | 85 | |||
XM_015145905.2 | XP_015001391.1 | 856 | 85 | |||
XM_028831210.1 | XP_028687043.1 | 875 | 85 | |||
GenBank mRNA | JU321232.1 | - | 787 | 85 | ||
JU473295.1 | - | 797 | 85 | |||
JV048550.1 | - | 711 | 85 | |||
JV452729.1 | - | 810 | 85 | |||
JV489142.1 | - | 780 | 85 | |||
JV704658.1 | - | 821 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |