Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105702.4 | XP_001105702.1 | 578 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |