Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085459.4 | XP_001085459.3 | 479 | 81 | ||
XM_015123478.2 | XP_014978964.2 | 132 | 81 | |||
GenBank mRNA | JV664417.1 | - | 151 | 81 | ||
JV699930.1 | - | 129 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |