Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102839.4 | XP_001102839.1 | 1470 | 110 | ||
GenBank mRNA | JU470817.1 | - | 1475 | 110 | ||
JU474573.1 | - | 1469 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |