Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266993.1 | NP_001253922.1 | 279 | 92 | ||
XM_015135410.2 | XP_014990896.1 | 281 | 92 | |||
XM_015135411.2 | XP_014990897.1 | 399 | 92 | |||
XM_015135412.2 | XP_014990898.1 | 471 | 92 | |||
GenBank mRNA | JU472759.1 | - | 214 | 92 | ||
JV043268.1 | - | 279 | 92 | |||
JV653736.1 | - | 174 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |