Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258120.1 | NP_001245049.1 | 193 | 149 | ||
GenBank mRNA | JU328615.1 | - | 110 | 149 | ||
JU473473.1 | - | 193 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |